  • Search Results
  • Detection of a broad range of Leishmania species and determination of parasite load of infected mouse by real-time PCR targeting the arginine permease gene AAP3.
Detection of a broad range of Leishmania species and determination of parasite load of infected mouse by real-time PCR targeting the arginine permease gene AAP3.
Acta tropica (2014-05-27)
Marit Gjerde Tellevik, Karl Erik Muller, Karen Rebbestad Løkken, Audun Helge Nerland
ABSTRACT
Leishmaniasis is one of the world's most neglected infectious diseases, affecting around 12 million people and more than 350 million at risk of infection. The clinical picture varies from self-healing cutaneous lesions to severe visceral infections, but still no commercial vaccines for humans are available and the currently used drugs have unpleasant side effects. Here we report a real-time PCR assay targeting the arginine permease gene AAP3 that can be applied for all the nine different species of the Leishmania genus tested; 4 Old World species and 5 New World species, from both L. (Leishmania) and L. (Viannia) subgenera. No cross-reaction was seen with Trypanosoma cruzi, Trypanosoma brucei, human or mouse genomic DNA. The assay has a high sensitivity, with a limit of detection of 10fg DNA for L. (L.) major and L. (L.) donovani, and 100fg DNA for L. (V.) braziliensis, and can be used for both qualitative and quantitative purposes. This AAP3-Assay, run in duplex with a host specific gene-assay, was also successfully used for quantification of parasite load of footpads from L. (L.) major-infected mice. It can therefore be a valuable tool in applications like monitoring effects of drugs, the selection of vaccine candidates and in screening patients, including asymptomatic carriers.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
Potassium chloride solution, for Ag/AgCl electrodes, ~3 M KCl, saturated with silver chloride
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%