  • Search Results
  • Concomitant RAS, RET/PTC, or BRAF mutations in advanced stage of papillary thyroid carcinoma.
Concomitant RAS, RET/PTC, or BRAF mutations in advanced stage of papillary thyroid carcinoma.
Thyroid : official journal of the American Thyroid Association (2014-05-07)
Minjing Zou, Essa Y Baitei, Ali S Alzahrani, Faisal S BinHumaid, Dania Alkhafaji, Roua A Al-Rijjal, Brian F Meyer, Yufei Shi
ABSTRACT
RET/PTC rearrangement, RAS, and BRAF mutations are considered to be mutually exclusive in papillary thyroid carcinoma (PTC). However, although concomitant mutations of RET/PTC, RAS, or BRAF have been reported recently, their significance for tumor progression and survival remains unclear. We sought to examine the prognostic value of concomitant mutations in PTC. We investigated 88 PTC for concomitant mutations. Mutation in BRAF exon 15, KRAS, NRAS, and HRAS were studied by polymerase chain reaction (PCR)-sequencing of tumor DNA; RET/PTC rearrangement was determined by reverse transcription (RT)-PCR-sequencing of tumor cDNA. BRAF(V600E) was detected in 39 of 82 classic PTC (CPTC) and in all three tall-cell variants (49%, 42/85). KRAS mutation (p.Q61R and p.S65N) was detected in two CPTC (2%, 2/88) and NRAS(Q61R) in one CPTC and two follicular variant PTC (FVPTC; 3%, 3/88). KRAS(S65N) was identified for the first time in thyroid cancer and could activate mitogen-associated protein kinase (MAPK). RET/PTC-1 was detected in nine CPTC, one tall-cell variant, and two FVPTC. Concomitant BRAF(V600E) and KRAS, or BRAF(V600E) and RET/PTC-1 mutations were found in two CPTC, and six CPTC and one tall-cell variant, respectively. In total, 11 concomitant mutations were found in 88 PTC samples (13%), and most of them were in the advanced stage of disease (8/11, 73%; p<0.01). Our data show that concomitant mutations are a frequent event in advanced PTC and are associated with poor prognosis. The concomitant mutations may represent intratumor heterogeneity and could exert a gene dosage effect to promote disease progression. KRAS(S65N) can constitutively activate the MAPK pathway.
Sigma-Aldrich
Anti-phospho-MAP2K1 (pThr386) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-MAP2K1 (pSer221) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-MAP2K1 (pThr291) antibody produced in rabbit, affinity isolated antibody