  • Search Results
  • CD90(+) human dermal stromal cells are potent inducers of FoxP3(+) regulatory T cells.
CD90(+) human dermal stromal cells are potent inducers of FoxP3(+) regulatory T cells.
The Journal of investigative dermatology (2014-07-23)
Karin Pfisterer, Karoline M Lipnik, Erhard Hofer, Adelheid Elbe-Bürger
ABSTRACT
The skin has to effectively combat external attacks, while maintaining skin immune homeostasis under steady-state conditions. To fulfill these challenging tasks, the dermis harbors a variety of heterogeneous cell types that are able to suppress T-cell proliferation similar to bone marrow mesenchymal stromal cells. Here we show that plastic-adherent, human dermal cells induce FoxP3 expression in TCR-complex-stimulated CD25(-)CD4(+)CD45RA(+) T cells in the absence of CD28 co-ligation in a cell-contact-dependent manner. These FoxP3(+) T cells reveal an effective suppressive capacity in vitro. Moreover, we found that the vast majority of CD90(+) dermal cells are perivascularly located and generate a significantly higher percentage of regulatory T cells compared with cells expressing markers such as CD271 in vitro. Importantly, we further demonstrate that plastic-adherent dermal cells are also able to differentiate toward the endothelial lineage. Our data show that human skin harbors specific cell types with immunosuppressive potential, which are located in close vicinity to their likely operational area and provide evidence for a CD28-independent regulatory mechanism. Further, the differentiation potential into endothelial cells suggests the existence of a tissue-resident cell pool for vessel regeneration. These findings might have important implications for the clinical use of allogeneic dermal cells to rebuild an imbalanced human skin immune homeostasis.
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle - FITC antibody produced in mouse, clone 1A4, purified immunoglobulin, buffered aqueous solution
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Indomethacin, meets USP testing specifications
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
Indomethacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Aphidicolin, analytical standard
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C