  • Search Results
  • Egr1 protein acts downstream of estrogen-leukemia inhibitory factor (LIF)-STAT3 pathway and plays a role during implantation through targeting Wnt4.
Egr1 protein acts downstream of estrogen-leukemia inhibitory factor (LIF)-STAT3 pathway and plays a role during implantation through targeting Wnt4.
The Journal of biological chemistry (2014-07-12)
Xiao-Huan Liang, Wen-Bo Deng, Ming Li, Zhen-Ao Zhao, Tong-Song Wang, Xu-Hui Feng, Yu-Jing Cao, En-Kui Duan, Zeng-Ming Yang
ABSTRACT
Embryo implantation is a highly synchronized process between an activated blastocyst and a receptive uterus. Successful implantation relies on the dynamic interplay of estrogen and progesterone, but the key mediators underlying embryo implantation are not fully understood. Here we show that transcription factor early growth response 1 (Egr1) is regulated by estrogen as a downstream target through leukemia inhibitory factor (LIF) signal transducer and activator of transcription 3 (STAT3) pathway in mouse uterus. Egr1 is localized in the subluminal stromal cells surrounding the implanting embryo on day 5 of pregnancy. Estrogen rapidly, markedly, and transiently enhances Egr1 expression in uterine stromal cells, which fails in estrogen receptor α knock-out mouse uteri. STAT3 is phosphorylated by LIF and subsequently recruited on Egr1 promoter to induce its expression. Our results of Egr1 expression under induced decidualization in vivo and in vitro show that Egr1 is rapidly induced after deciduogenic stimulus. Egr1 knockdown can inhibit in vitro decidualization of cultured uterine stromal cells. Chromatin immunoprecipitation data show that Egr1 is recruited to the promoter of wingless-related murine mammary tumor virus integration site 4 (Wnt4). Collectively, our study presents for the first time that estrogen regulates Egr1 expression through LIF-STAT3 signaling pathway in mouse uterus, and Egr1 functions as a critical mediator of stromal cell decidualization by regulating Wnt4.
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Tyrosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Supelco
Digoxigenin, analytical standard
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
DL-Tyrosine, 99%
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Supelco
Progesterone, VETRANAL®, analytical standard
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
Progesterone, European Pharmacopoeia (EP) Reference Standard
Digoxigenin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
Magnesium chloride solution, 0.1 M