BCR-ABL affects STAT5A and STAT5B differentially.
PloS one (2014-05-20)
Michael Schaller-Schönitz, David Barzan, Andrew J K Williamson, John R Griffiths, Iris Dallmann, Karin Battmer, Arnold Ganser, Anthony D Whetton, Michaela Scherr, Matthias Eder
ABSTRACT
Signal transducers and activators of transcription (STATs) are latent cytoplasmic transcription factors linking extracellular signals to target gene transcription. Hematopoietic cells express two highly conserved STAT5-isoforms (STAT5A/STAT5B), and STAT5 is directly activated by JAK2 downstream of several cytokine receptors and the oncogenic BCR-ABL tyrosine kinase. Using an IL-3-dependent cell line with inducible BCR-ABL-expression we compared STAT5-activation by IL-3 and BCR-ABL in a STAT5-isoform specific manner. RNAi targeting of STAT5B strongly inhibits BCR-ABL-dependent cell proliferation, and STAT5B but not STAT5A is essential for BCL-XL-expression in the presence of BCR-ABL. Although BCR-ABL induces STAT5-tyrosine phosphorylation independent of JAK2-kinase activity, BCR-ABL is less efficient in inducing active STAT5A:STAT5B-heterodimerization than IL-3, leaving constitutive STAT5A and STAT5B-homodimerization unaffected. In comparison to IL-3, nuclear accumulation of a STAT5A-eGFP fusion protein is reduced by BCR-ABL, and BCR-ABL tyrosine kinase activity induces STAT5A-eGFP translocation to the cell membrane and co-localization with the IL-3 receptor. Furthermore, BCR-ABL-dependent phosphorylation of Y682 in STAT5A was detected by mass-spectrometry. Finally, RNAi targeting STAT5B but not STAT5A sensitizes human BCR-ABL-positive cell lines to imatinib-treatment. These data demonstrate differences between IL-3 and BCR-ABL-mediated STAT5-activation and isoform-specific effects, indicating therapeutic options for isoform-specific STAT5-inhibition in BCR-ABL-positive leukemia.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
1,4-Diazabicyclo[2.2.2]octane, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride, ≥97.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Dabco® 33-LV
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Pefabloc® SC, analytical standard
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)