  • Search Results
  • Monitoring of epidermal growth factor receptor tyrosine kinase inhibitor-sensitizing and resistance mutations in the plasma DNA of patients with advanced non-small cell lung cancer during treatment with erlotinib.
Monitoring of epidermal growth factor receptor tyrosine kinase inhibitor-sensitizing and resistance mutations in the plasma DNA of patients with advanced non-small cell lung cancer during treatment with erlotinib.
Cancer (2014-08-12)
Boe S Sorensen, Lin Wu, Wen Wei, Julie Tsai, Britta Weber, Ebba Nexo, Peter Meldgaard
ABSTRACT
The feasibility of monitoring epidermal growth factor receptor (EGFR) mutations in plasma DNA from patients with advanced non-small cell lung cancer (NSCLC) during treatment with erlotinib and its relation to disease progression was investigated. The amount of EGFR-mutant DNA was tested in plasma DNA from patients with advanced NSCLC with allele-specific polymerase chain reaction assays. Blood samples from 23 patients with adenocarcinoma of NSCLC that carried tyrosine kinase inhibitor-sensitizing EGFR mutations were taken immediately before treatment with erlotinib. Additional blood samples were taken at timed intervals until erlotinib treatment was withdrawn. The amount of plasma DNA with sensitizing EGFR mutations was found to be reduced after the first cycle of erlotinib treatment in 22 of 23 patients (96%). No patients presented with the resistant T790M mutation in the pretreatment sample, but at the time of disease progression the mutation was detected in plasma from 9 patients (39%). The quantitative data from the current study demonstrated that when a T790M mutation emerged in the blood it was accompanied by an increase in the original sensitizing EGFR mutation. When T790M was detected, it was found to be present in all subsequent blood samples from that patient. Most interestingly, the results of the current study demonstrated that monitoring the EGFR mutations in the blood allows for the detection of the T790M mutation up to 344 days before disease progression is clinically evident (range, 15-344 days). The results of the current study demonstrated that serial monitoring of EGFR mutations in plasma DNA is feasible and may allow for the early detection of resistance mutations. These results warrant further studies to explore the clinical usefulness of such analysis.
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Supelco
2-Propanol, analytical standard
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade