  • Search Results
  • Sperm characteristics, antioxidant status and hormonal profile in rats treated with artemisinin.
Sperm characteristics, antioxidant status and hormonal profile in rats treated with artemisinin.
Andrologia (2013-10-02)
E O Farombi, I A Adedara, A O Abolaji, J P Anamelechi, J O Sangodele
ABSTRACT
The indiscriminate use, abuse and patients' noncompliance to normal prescription of artemisinin and its derivatives are a common practice during the treatment for drug-resistant malaria parasites in most developing countries. This study investigated the influence of artemisinin on the testicular and epididymal sperm antioxidant systems as well as on the plasma levels of hormones from the pituitary and thyroid components of the brain-pituitary-testicular axis. Oral exposure of rats to 0, 7 and 35 mg kg(-1) artemisinin for 7 days showed that the testicular antioxidant status at both therapeutic dose (7 mg kg(-1) ) and overdose (35 mg kg(-1) ), and the sperm antioxidant status at therapeutic dose of artemisinin remained unaffected compared with control. However, increased hydrogen peroxide and lipid peroxidation levels were accompanied by a concomitant decrease in glutathione peroxidase and glutathione-S-transferase activities as well as glutathione level in spermatozoon of rats administered with overdose of artemisinin. While plasma levels of all the hormones investigated remained unaffected, severe epididymal degeneration with concomitant decrease in sperm quantity and quality was observed in rats treated with overdose of artemisinin compared with control. Overall, induction of oxidative stress in the epididymis, but not in the testes, could cause reproductive deficits in individuals unduly undergoing artemisinin therapy.
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, 97%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Supelco
Trizma® base, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O