  • Search Results
  • Visualization of HTLV-1-specific cytotoxic T lymphocytes in the spinal cords of patients with HTLV-1-associated myelopathy/tropical spastic paraparesis.
Visualization of HTLV-1-specific cytotoxic T lymphocytes in the spinal cords of patients with HTLV-1-associated myelopathy/tropical spastic paraparesis.
Journal of neuropathology and experimental neurology (2014-12-04)
Eiji Matsuura, Ryuji Kubota, Yuetsu Tanaka, Hiroshi Takashima, Shuji Izumo
ABSTRACT
Activated human T-lymphotropic virus type-1 (HTLV-1)-specific CD8-positive cytotoxic T lymphocytes (CTLs) are markedly increased in the periphery of patients with HTLV-1-associated myelopathy/tropical spastic paraparesis (HAM/TSP), an HTLV-1-induced inflammatory disease of the CNS. Although virus-specific CTLs play a pivotal role to eliminate virus-infected cells, the potential role of HTLV-1-specific CTLs in the pathogenesis of HAM/TSP remains unclear. To address this issue, we evaluated the infiltration of HTLV-1-specific CTLs and the expression of HTLV-1 proteins in the spinal cords of 3 patients with HAM/TSP. Confocal laser scanning microscopy with our unique staining procedure made it possible to visualize HTLV-1-specific CTLs infiltrating the CNS of the HAM/TSP patients. The frequency of HTLV-1-specific CTLs was more than 20% of CD8-positive cells infiltrating the CNS. In addition, HTLV-1 proteins were detected in CD4-positive infiltrating T lymphocytes but not CNS resident cells. Although neurons were generally preserved, apoptotic oligodendrocytes were frequently in contact with CD8-positive cells; this likely resulted in demyelination. These findings suggest that the immune responses of the CTLs against HTLV-1-infected CD4-positive lymphocytes migrating into the CNS resulted in bystander neural damage.
Product Description
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.