  • Search Results
  • Quantitative targeted absolute proteomics for 28 human transporters in plasma membrane of Caco-2 cell monolayer cultured for 2, 3, and 4 weeks.
Quantitative targeted absolute proteomics for 28 human transporters in plasma membrane of Caco-2 cell monolayer cultured for 2, 3, and 4 weeks.
Drug metabolism and pharmacokinetics (2015-03-15)
Yasuo Uchida, Sumio Ohtsuki, Junichi Kamiie, Ken Ohmine, Ryo Iwase, Tetsuya Terasaki
ABSTRACT
The purpose of the present study was to evaluate and compare the absolute protein expression levels of 28 drug-related transporters in Caco-2 cell monolayers cultured for 2, 3, and 4 weeks. Plasma membrane fractions of Caco-2 cells cultured on Transwell inserts for 2, 3 and 4 weeks were prepared and digested with trypsin, and then simultaneous absolute quantification of 28 transporters and Na(+)/K(+)-ATPase was conducted using our established quantitative targeted absolute proteomic technique. Nine transporters and Na(+)/K(+)-ATPase were detected. MDR1, BCRP, PEPT1, OSTα and OSTβ were highly expressed (greater than 1 fmol/μg protein), while MRP2, MRP4, OATP2B1 and MCT1 were moderately expressed (0.328-0.871 fmol/μg protein). No significant difference was observed in the protein expression levels of these transporters or Na(+)/K(+)-ATPase among the 2-, 3- and 4-week cultures. The other 19 transporters, including MRP1, MRP3, OATP1A2, OATP3A1, OATP4A1, and OATP1B3, were not detected. This information about the rank order of transporter protein expression will be useful to predict what transporter(s) are likely or unlikely to influence the permeability of various compounds across monolayers of Caco-2 cells, which are widely used in drug development studies.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Sucrose, ≥99.5%
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
SAFC
L-Glutamine
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)