  • Search Results
  • Biodegradable polymeric coatings on cochlear implant surfaces and their influence on spiral ganglion cell survival.
Biodegradable polymeric coatings on cochlear implant surfaces and their influence on spiral ganglion cell survival.
Journal of biomedical materials research. Part B, Applied biomaterials (2014-01-18)
P Ceschi, A Bohl, K Sternberg, A Neumeister, V Senz, K P Schmitz, M Kietzmann, V Scheper, T Lenarz, T Stöver, G Paasche
ABSTRACT
To improve the electrode-nerve interface of cochlear implants (CI), the role of poly(L-lactide) (PLLA) and poly(4-hydroxybutyrate) (P(4HB)) as potential coating matrices for CI was assessed both in vitro and in vivo in terms of degradation behavior and effects on spiral ganglion neurons, the main target of the electrical stimulation with a CI. Growth rates of fibroblasts on the polymers were investigated and a direct-contact test with freshly isolated spiral ganglion cells (SGC) was performed. In addition, the effects of the polymer degradation inside the inner ear were evaluated in vivo. The polymer degradation was assessed by use of scanning electron microscopy in combination with an energy-dispersive X-ray analysis. In vitro, no influence of the polymers was detected on fibroblasts' viability and on SGC survival rate. In vivo, SGC density was decreased only 6 months after implantation in the basal and middle turns of the cochlea in comparison to normal-hearing animals but not between implanted groups (coated or uncoated). The analysis of the electrode models showed that in vivo P(4HB) is characterized by a gradual degradation completed after 6 months; whereas, the PLLA coatings burst along their longitudinal axis but showed only little degradation within the same time frame. In conclusion, both polymers seem to justify further evaluation as possible coating for CI electrodes. Of the two options, due to its excellent coating adhesion/stability and optimal degradation behavior, P(4HB) may prove to be the more promising biodegradable polymer for designing a drug delivery system from the surface of CI electrodes.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
SAFC
HEPES
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Trypan Blue solution, 0.4%, for microscopy
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
SAFC
HEPES