  • Search Results
  • Case Study of a γ-Butyrolactone Alkylation with 1,3-Dimethyl-2-imidazolidinone as a Promoter.
Case Study of a γ-Butyrolactone Alkylation with 1,3-Dimethyl-2-imidazolidinone as a Promoter.
Organic Process Research & Development (2001)
Li B, et al.