  • Search Results
  • The WNT2 gene polymorphism associated with speech delay inherent to autism.
The WNT2 gene polymorphism associated with speech delay inherent to autism.
Research in developmental disabilities (2012-04-24)
Ping-I Lin, Yi-Ling Chien, Yu-Yu Wu, Chia-Hsiang Chen, Susan Shur-Fen Gau, Yu-Shu Huang, Shih-Kai Liu, Wen-Che Tsai, Yen-Nan Chiu
ABSTRACT
Previous evidence suggests that language function is modulated by genetic variants on chromosome 7q31-36. However, it is unclear whether this region harbors loci that contribute to speech delay in autism. We previously reported that the WNT2 gene located on 7q31 was associated with the risk of autism. Additionally, two other genes on 7q31-36, FOXP2 and the EN2 genes are also found to play a role in language impairment. Therefore, we hypothesize that the WNT2 gene, FOXP2 gene, and EN2 gene, may act in concert to influence language development in the same population. A total of 373 individuals diagnosed with autistic disorder were recruited in the current study. We selected 6 tag single nucleotide polymorphisms (SNPs) within the WNT2 gene, 3 tag SNPs in the FOXP2, and 3 tag SNPs in the EN2 genes, to study the effect of these genes on language development. Age of first phrase was treated as a quantitative trait. We used general linear model to assess the association between speech delay and these variants. The results show that rs2896218 in the WNT2 gene was moderately significantly associated with age of first phrase (permutation p = 0.0045). A three-locus haplotype in the WNT2 gene was significantly associated with age of first phrase (permutation p = 2 × 10(-4)). Furthermore, we detected an interaction effect on age of first phrase between a SNP rs2228946 in the WNT2 gene and another SNP rs6460013 in the EN2 gene (p = 0.0012). Therefore, the WNT2 gene may play a suggestive role in language development in autistic disorder. Additionally, the WNT2 gene and EN2 gene may act in concert to influence the language development in autism.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.