  • Search Results
  • Chronic helminth infection and helminth-derived egg antigens promote adipose tissue M2 macrophages and improve insulin sensitivity in obese mice.
Chronic helminth infection and helminth-derived egg antigens promote adipose tissue M2 macrophages and improve insulin sensitivity in obese mice.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2015-04-09)
Leonie Hussaarts, Noemí García-Tardón, Lianne van Beek, Mattijs M Heemskerk, Simone Haeberlein, Gerard C van der Zon, Arifa Ozir-Fazalalikhan, Jimmy F P Berbée, Ko Willems van Dijk, Vanessa van Harmelen, Maria Yazdanbakhsh, Bruno Guigas
ABSTRACT
Chronic low-grade inflammation associated with obesity contributes to insulin resistance and type 2 diabetes. Helminth parasites are the strongest natural inducers of type 2 immune responses, and short-lived infection with rodent nematodes was reported to improve glucose tolerance in obese mice. Here, we investigated the effects of chronic infection (12 weeks) with Schistosoma mansoni, a helminth that infects millions of humans worldwide, on whole-body metabolic homeostasis and white adipose tissue (WAT) immune cell composition in high-fat diet-induced obese C57BL/6 male mice. Our data indicate that chronic helminth infection reduced body weight gain (-62%), fat mass gain (-89%), and adipocyte size; lowered whole-body insulin resistance (-23%) and glucose intolerance (-16%); and improved peripheral glucose uptake (+25%) and WAT insulin sensitivity. Analysis of immune cell composition by flow cytometry and quantitative PCR (qPCR) revealed that S. mansoni promoted strong increases in WAT eosinophils and alternatively activated (M2) macrophages. Importantly, injections with S. mansoni-soluble egg antigens (SEA) recapitulated the beneficial effect of parasite infection on whole-body metabolic homeostasis and induced type 2 immune responses in WAT and liver. Taken together, we provide novel data suggesting that chronic helminth infection and helminth-derived molecules protect against metabolic disorders by promoting a T helper 2 (Th2) response, eosinophilia, and WAT M2 polarization.
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
SAFC
HEPES
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)