Printed paper-based arrays as substrates for biofilm formation.
AMB Express (2014-07-10)
Anni Määttänen, Adyary Fallarero, Janni Kujala, Petri Ihalainen, Pia Vuorela, Jouko Peltonen
ABSTRACT
The suitability of paper-based arrays for biofilm formation studies by Staphylococcus aureus is demonstrated. Laboratory-coated papers with different physicochemical properties were used as substrates. The array platform was fabricated by patterning the coated papers with vinyl-substituted polydimethylsiloxane (PDMS) -based ink. The affinity of bacteria onto the flexographically printed hydrophobic and smooth PDMS film was very low whereas bacterial adhesion and biofilm formation occurred preferentially on the unprinted areas, i.e. in the reaction arrays. The concentration of the attached bacteria was quantified by determining the viable colony forming unit (CFU/cm(2)) numbers. The distribution and the extent of surface coverage of the biofilms were determined by atomic force microscopy. In static conditions, the highest bacterial concentration and most highly organized biofilms were observed on substrates with high polarity. On a rough paper surface with low polarity, the biofilm formation was most hindered. Biofilms were effectively removed from a polar substrate upon exposure to (+)-dehydroabietic acid, an anti-biofilm compound.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Pyridinium chlorochromate, 98%
Sigma-Aldrich
Calcium carbonate, ACS reagent, chelometric standard, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%