  • Search Results
  • Determination of BMAA and three alkaloid cyanotoxins in lake water using dansyl chloride derivatization and high-resolution mass spectrometry.
Determination of BMAA and three alkaloid cyanotoxins in lake water using dansyl chloride derivatization and high-resolution mass spectrometry.
Analytical and bioanalytical chemistry (2015-05-04)
Audrey Roy-Lachapelle, Morgan Solliec, Sébastien Sauvé
ABSTRACT
A new analytical method was developed for the detection of alkaloid cyanotoxins in harmful algal blooms. The detection of the nonproteinogenic amino acid β-N-methylamino-L-alanine (BMAA) and two of its conformation isomers, 2,4-diaminobutyric acid (DAB) and N-(2-aminoethyl) glycine (AEG), as well as three alkaloid cyanotoxins, anatoxin-a (ANA-a), cylindrospermopsin (CYN), and saxitoxin (STX), is presented. The use of a chemical derivatization with dansyl chloride (DNS) allows easier separation with reversed phase liquid chromatography. Detection with high-resolution mass spectrometry (HRMS) with the Q-Exactive enables high selectivity with specific fragmentation as well as exact mass detection, reducing considerably the possibilities of isobaric interferences. Previous to analysis, a solid phase extraction (SPE) step is used for purification and preconcentration. After DNS derivatization, samples are submitted to ultra high-performance liquid chromatography coupled with heated electrospray ionisation and the Q-Exactive mass spectrometer (UHPLC-HESI-HRMS). With an internal calibration using isotopically-labeled DAB-D3, the method was validated with good linearity (R (2)  > 0.998), and method limits of detection and quantification (MLD and MLQ) for target compounds ranged from 0.007 to 0.01 μg L(-1) and from 0.02 to 0.04 μg L(-1), respectively. Accuracy and within-day/between-days variation coefficients were below 15%. SPE recovery values ranged between 86 and 103%, and matrix effects recovery values ranged between 75 and 96%. The developed analytical method was successfully validated with 12 different lakes samples, and concentrations were found ranging between 0.009 and 0.3 μg L(-1) except for STX which was not found in any sample.
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
DL-Phenylalanine, ReagentPlus®, 99%
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Ethylene glycol monophenyl ether, ≥90%
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium tetraborate, 99.998% trace metals basis
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Sodium tetraborate, 99%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Dansyl chloride, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
SAFC
Glycine
Sigma-Aldrich
3,5-Dinitrosalicylic acid, used in colorimetric determination of reducing sugars
Sigma-Aldrich
3,5-Dinitrosalicylic acid, 98%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%