  • Search Results
  • VE1 immunohistochemistry predicts BRAF V600E mutation status and clinical outcome in colorectal cancer.
VE1 immunohistochemistry predicts BRAF V600E mutation status and clinical outcome in colorectal cancer.
Oncotarget (2015-10-27)
Christian Schafroth, José A Galván, Irene Centeno, Viktor H Koelzer, Heather E Dawson, Lena Sokol, Gregor Rieger, Martin D Berger, Marion Hädrich, Robert Rosenberg, Ulrich Nitsche, Beat Schnüriger, Rupert Langer, Daniel Inderbitzin, Alessandro Lugli, Inti Zlobec
ABSTRACT
VE1 is a monoclonal antibody detecting mutant BRAFV(600E) protein by immunohistochemistry. Here we aim to determine the inter-observer agreement and concordance of VE1 with mutational status, investigate heterogeneity in colorectal cancers and metastases and determine the prognostic effect of VE1 in colorectal cancer patients. Concordance of VE1 with mutational status and inter-observer agreement were tested on a pilot cohort of colorectal cancers (n = 34), melanomas (n = 23) and thyroid cancers (n = 8). Two prognostic cohorts were evaluated (n = 259, Cohort 1 and n = 226, Cohort 2) by multiple-punch tissue microarrays. VE1 staining on preoperative biopsies (n = 118 patients) was compared to expression in resections. Primary tumors and metastases from 13 patients were tested for VE1 heterogeneity using a tissue microarray generated from all available blocks (n = 100 blocks). Inter-observer agreement was 100% (kappa = 1.0). Concordance between VE1 and V600E mutation was 98.5%. Cohort 1: VE1 positivity (seen in 13.5%) was associated with older age (p = 0.0175) and MLH1 deficiency (p < 0.0001). Cohort 2: VE1 positivity (seen in 12.8%) was associated with female gender (p = 0.0016), right-sided tumor location (p < 0.0001), higher tumor grade (p < 0.0001) and mismatch repair (MMR)-deficiency (p < 0.0001). In survival analysis, MMR status and postoperative therapy were identified as possible confounding factors. Adjusting for these features, VE1 was an unfavorable prognostic factor. Preoperative biopsy staining matched resections in all cases except one. No heterogeneity was found across any primary/metastatic tumor blocks. VE1 is highly concordant for V600E and homogeneously expressed suggesting staining can be analysed on resection specimens, preoperative biopsies, metastatic lesions and tissue microarrays.
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Supelco
Hematoxylin
Sigma-Aldrich
Trimesic acid, 95%
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Trimethylaluminum, 97%
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in toluene
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in hexanes
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Trimethylaluminum, packaged for use in deposition systems
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in heptane