  • Search Results
  • Dopamine-sensitive signaling mediators modulate psychostimulant-induced ultrasonic vocalization behavior in rats.
Dopamine-sensitive signaling mediators modulate psychostimulant-induced ultrasonic vocalization behavior in rats.
Behavioural brain research (2015-08-16)
Stacey N Williams, Ashiwel S Undieh
ABSTRACT
The mesolimbic dopamine system plays a major role in psychostimulant-induced ultrasonic vocalization (USV) behavior in rodents. Within this system, psychostimulants elevate synaptic concentrations of dopamine thereby leading to exaggerated activation of postsynaptic dopamine receptors within the D1-like and D2-like subfamilies. Dopamine receptor stimulation activate several transmembrane signaling systems and cognate intracellular mediators; downstream activation of transcription factors then conveys the information from receptor activation to appropriate modulation of cellular and physiologic functions. We previously showed that cocaine-induced USV behavior was associated with enhanced expression of the neurotrophin BDNF. Like cocaine, amphetamine also increases synaptic dopamine levels, albeit primarily through facilitating dopamine release. Therefore, in the present study we investigated whether amphetamine and cocaine similarly activate dopamine-linked signaling cascades to regulate intracellular mediators leading to induction of USV behavior. The results show that amphetamine increased the emission of 50 kHz USVs and this effect was blocked by SCH23390, a D1 receptor antagonist. Similar to cocaine, amphetamine increased BDNF protein expression in discrete brain regions, while pretreatment with K252a, a trkB neurotrophin receptor inhibitor, significantly reduced amphetamine-induced USV behavior. Inhibition of cyclic-AMP/PKA signaling with H89 or inhibition of PLC signaling with U73122 significantly blocked both the acute and subchronic amphetamine-induced USV behavior. In contrast, pharmacologic inhibition of either pathway enhanced cocaine-induced USV behavior. Although cocaine and amphetamine similarly modulate neurotrophin expression and USV, the molecular mechanisms by which these psychostimulants differentially activate dopamine receptor subtypes or other monoaminergic systems may be responsible for the distinct aspects of behavioral responses.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
K-252a, from Nocardiopsis sp., ≥96.0% (HPLC)
Sigma-Aldrich
K-252a, Ready Made Solution, from Nonomuraea longicatena, >98%
Sigma-Aldrich
K-252a, from Nocardiopsis sp.
Sigma-Aldrich
K-252a, from Nonomuraea longicatena, ≥98% (HPLC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E422, anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture