  • Search Results
  • Neuroprotective Effects of (-)-Epigallocatechin-3-Gallate Against Focal Cerebral Ischemia/Reperfusion Injury in Rats Through Attenuation of Inflammation.
Neuroprotective Effects of (-)-Epigallocatechin-3-Gallate Against Focal Cerebral Ischemia/Reperfusion Injury in Rats Through Attenuation of Inflammation.
Neurochemical research (2015-07-23)
FengJin Zhang, Na Li, LinLan Jiang, LongHao Chen, MuTu Huang
ABSTRACT
Stroke is the second leading cause of death among adults worldwide. (-)-Epigallocatechin-3-gallate (EGCG) has been demonstrated to exhibit neuroprotective functions in cerebral ischemia/reperfusion injury. However, the underlying mechanisms in this process and its contribution to the protection function remain unknown. The current study examined the neuroprotective effects of EGCG after transient middle cerebral artery occlusion (tMCAO) in rats. tMCAO for 120 min was induced in male Sprague-Dawley rats treated with EGCG (50 mg/kg, i.p.) or Vehicle immediately after reperfusion. Neurological score, infarct ratio and inflammation-related molecules (assessed by 2,3,5-triphenyltetrazolium chloride, enzyme-linked immunosorbent assays, quantitative real-time PCR or western blotting) were estimated at 24 h after operation. EGCG prevented the impairment of neurological function and decreased the infarct volume, compared with the Vehicle group. The inflammation-related molecules TNF-α, IL-1β, IL-6 levels usually caused by ischemia/reperfusion were significantly ameliorated by EGCG. EGCG also inhibited the upregulation of nuclear factor-kappa B/p65 (NF-κB/p65), and induction of cyclooxygenase 2 and inducible nitric oxide synthase. The present study indicates that EGCG may be a promising therapeutic agent for cerebral ischemia/reperfusion injury through attenuation of inflammation.
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
Titanium tetrachloride, packaged for use in deposition systems