  • Search Results
  • A novel method for the rapid determination of polyethoxylated tallow amine surfactants in water and sediment using large volume injection with high performance liquid chromatography and tandem mass spectrometry.
A novel method for the rapid determination of polyethoxylated tallow amine surfactants in water and sediment using large volume injection with high performance liquid chromatography and tandem mass spectrometry.
Analytica chimica acta (2015-09-08)
Andrew R S Ross, Xiangjun Liao
ABSTRACT
Polyethoxylated tallow amine (POEA) surfactants have been used in many glyphosate-based herbicide formulations for agricultural, industrial and residential weed control. The potential for release of these compounds into the environment is of increasing concern due to their toxicity towards aquatic organisms. Current methods for analysis of POEA surfactants require significant time and effort to achieve limits of quantification that are often higher than the concentrations at which biological effects have been observed (as low as 2 ng mL(-1)). We have developed a rapid and robust method for quantifying the POEA surfactant mixture MON 0818 at biologically relevant concentrations in fresh water, sea water and lake sediment using reversed phase high-performance liquid chromatography and electrospray ionization-tandem mass spectrometry. Water samples preserved by 1:1 v/v dilution with methanol are analyzed directly following centrifugation. Sediment samples undergo accelerated solvent extraction in aqueous methanol prior to analysis. Large volume (100 μL) sample injection and multiple reaction monitoring of a subset of the most abundant POEA homologs provide limits of quantification of 0.5 and 2.9 ng mL(-1) for MON 0818 in fresh water and sea water, respectively, and 2.5 ng g(-1) for total MON 0818 in lake sediment. Average recoveries of 93 and 75% were achieved for samples of water and sediment, respectively spiked with known amounts of MON 0818. Precision and accuracy for the analysis of water and sediment samples were within 10 and 16%, respectively based upon replicate analyses of calibration standards and representative samples. Results demonstrate the utility of the method for quantifying undegraded MON 0818 in water and sediment, although a more comprehensive method may be needed to identify and determine other POEA mixtures and degradation profiles that might occur in the environment.
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles