  • Search Results
  • Acidified bile acids increase hTERT expression via c-myc activation in human gastric cancer cells.
Acidified bile acids increase hTERT expression via c-myc activation in human gastric cancer cells.
Oncology reports (2015-04-16)
Xiaolong Wang, Peihua Zhou, Xuejun Sun, Jianbao Zheng, Guangbing Wei, Li Zhang, Hui Wang, Jianfeng Yao, Shaoying Lu, Pengbo Jia
ABSTRACT
Human telomerase reverse transcriptase (hTERT) is upregulated in most cancer cell types as well in immortalized cells. The underlying mechanism for such upregulation, however, remains largely unknown. We report here that bile acids under acidified media increase hTERT expression via c-myc activation in primary human gastric cancer cell lines. Human gastric cancer MKN28, MGC803 and SGC7901 cells were treated with 100 µM deoxycholic acid (DCA) or chenodeoxycholic acid (CDCA) with or without acidified media in the presence or absence of the c-myc inhibitor 10058-F4 for 24 h. hTERT and c-myc protein levels were determined by western blot analysis. hTERT and c-myc mRNA levels were determined by RT-PCR. The promoter activities of hTERT and c-myc transcription were determined using promoter reporter luciferase assays for both. Telomerase enzyme activity was analyzed by stretch PCR. hTERT mRNA and protein levels were significantly increased by bile acids in acidified media and were accompanied with enhanced telomerase activity. No changes were found at a pH of 7.0 or with acidified media alone. Similarly, the mRNA and protein levels of c-myc were also increased by bile acids in acidified media but not at a pH of 7.0 or with acidified media alone. Importantly, pharmacologic inhibition of c-myc using 10058-F4 prevented hTERT induction by DCA or CDCA in gastric cancer cells under acidic conditions. Bile acids (DCA and CDCA) under acidic conditions increased hTERT expression in human gastric cancer cells by activation of c-myc transcription. This suggests that acidified bile acids may promote tumorigenesis and affect cell ageing via telomerase activation.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium pyrophosphate tetrabasic, ≥95%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Rhodanine, 97%
Sigma-Aldrich
Sodium fluoride solution
Sigma-Aldrich
10058-F4, ≥98% (HPLC), solid
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)