  • Search Results
  • Development of a freeze-dried kit formulation for the preparation of 99mTc-NTP 15-5, a radiotracer for scintigraphic imaging of proteoglycans.
Development of a freeze-dried kit formulation for the preparation of 99mTc-NTP 15-5, a radiotracer for scintigraphic imaging of proteoglycans.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2015-03-31)
Aurélien Vidal, Vincent Gaumet, Marie-Josèphe Galmier, Sophie Besse, Fernand Leal, Françoise Gachon, Gilles Viot, Jacques Métin, Jean-Michel Chezal, Elisabeth Miot-Noirault, Philippe Auzeloux
ABSTRACT
The cartilage-targeting strategy is based on the strong affinity of quaternary ammonium (QA) functions for cartilage proteoglycans. We use a bifunctional agent containing QA moiety and a polyazamacrocycle structure able to complex technetium-99m. (99m)Tc-NTP 15-5 was selected for its high stability and its high affinity for proteoglycans in vivo. Labeling conditions of NTP 15-5 were optimized, and a lyophilized kit was developed for radiolabeling of (99m)Tc-NTP 15-5 (radiochemical yields 94.6±1.8%). (99m)Tc-NTP 15-5 was stable and resulted in favorable biological evaluations.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium, cubes, contains mineral oil, 99.9% trace metals basis
Sigma-Aldrich
Sodium, in kerosene, pieces (large), ≥99.8% (sodium basis)
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%
Sigma-Aldrich
Tin(II) chloride dihydrate, ≥99.995% trace metals basis
Sigma-Aldrich
Sodium, ACS reagent, dry
Sigma-Aldrich
Sodium, 99.95% trace metals basis, ingot
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Sodium, 25-35 wt % dispersion in paraffin
Sigma-Aldrich
Tin(II) chloride dihydrate, ≥99.99% trace metals basis
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Supelco
Toluene, HPLC grade, 99.8%