  • Search Results
  • Silver nanoparticles anchored on nitrogen-doped graphene as a novel electrochemical biosensing platform with enhanced sensitivity for aptamer-based pesticide assay.
Silver nanoparticles anchored on nitrogen-doped graphene as a novel electrochemical biosensing platform with enhanced sensitivity for aptamer-based pesticide assay.
The Analyst (2015-08-08)
Ding Jiang, Xiaojiao Du, Qian Liu, Lei Zhou, Liming Dai, Jing Qian, Kun Wang
ABSTRACT
Silver nanoparticles (NPs) decorated nitrogen doped graphene (NG) nanocomposites were prepared through a one-step thermal-treatment route using arginine as the nitrogen source. By integrating the excellent electrical properties and large surface area of Ag NPs and NG, the obtained Ag/NG nanocomposites show more effective electron transfer and high loading capacity than Ag-graphene and pure NG. In the presence of a target, the stronger interaction between the aptamer and the target promotes the formation of a target-aptamer complex on the electrode surface which blocks the electron transfer. Based on this sensing mechanism, a novel and highly sensitive biosensing platform by the use of Ag/NG as enhancing materials is demonstrated for detection of the model target, acetamiprid. The presented aptasensor exhibited a wide linear response for acetamiprid in the range of 1 × 10(-13) M to 5 × 10(-9) M with a low detection limit of 3.3 × 10(-14) M (S/N = 3). Moreover, this electrochemical aptasensor avoided complicated labeling procedures and showed magnificent sensitivity, high selectivity and low cost, which made it not only convenient but also time-saving and applicable. Furthermore, the proposed design may offer a promising way to develop a new electrochemical aptasensor for sensitive and specific detection of a wide spectrum of analytes in food, medical and environmental fields.
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
6-Mercapto-1-hexanol, 97%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.