  • Search Results
  • A gold nanoparticle-based colorimetric probe for rapid detection of 1-hydroxypyrene in urine.
A gold nanoparticle-based colorimetric probe for rapid detection of 1-hydroxypyrene in urine.
The Analyst (2015-05-20)
Yaqi Hu, Chunyan Du, Yunchao Li, Louzhen Fan, Xiaohong Li
ABSTRACT
Direct and rapid detection of 1-hydroxypyrene (1-OHP) is of great importance owing to its high carcinogenicity, teratogenicity and toxicity. In this study, a simple colorimetric assay for rapid determination of 1-OHP is reported, which is based on non-crosslinking aggregation of gold nanoparticles (Au NPs) induced by 1-OHP in the presence of formic acid (FA). Initially, Au NPs were synthesized with citrate as the capping agent and exhibited red color. Subsequently, the addition of FA did not cause aggregation of Au NPs, but a proton transfer process occurred from FA to carboxylic anions on the surface of Au NPs with a decreased zeta potential. The subsequent addition of 1-OHP resulted in a further decreased zeta potential and an intensely hydrophobic environment, which led to a strong and rapid non-crosslinking aggregation of Au NPs within 5 min with the color changing from red to violet blue. Based on this principle, sensitive and selective detection of 1-OHP was achieved. The detection limit was 3.3 nM. Finally, the colorimetric assay was successfully applied to detect 1-OHP in a urine sample. This strategy provides new insights into developing colorimetric methods for on-site and real-time detection of polycyclic aromatic hydrocarbons.
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
Sigma-Aldrich
Uric acid, ≥99%, crystalline
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, meets USP testing specifications
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
1-Naphthol, ReagentPlus®, ≥99%
Sigma-Aldrich
Resorcinol, ACS reagent, ≥99.0%
Sigma-Aldrich
2-Naphthol, 99%
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
Sigma-Aldrich
Resorcinol, ReagentPlus®, 99%
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Urea, BioUltra, for molecular biology, ≥99% (T)