  • Search Results
  • The components of the angiotensin-(1-7) system are differentially expressed during follicular wave in cattle.
The components of the angiotensin-(1-7) system are differentially expressed during follicular wave in cattle.
Journal of the renin-angiotensin-aldosterone system : JRAAS (2013-06-15)
Marcos Henrique Barreta, Bernardo Garziera Gasperin, Rogério Ferreira, Monique Rovani, Gabriel Ribas Pereira, Rodrigo C Bohrer, João Francisco de Oliveira, Paulo Bayard Dias Gonçalves
ABSTRACT
This study was based on the hypothesis that some components of the angiotensin-(1-7) (Ang-(1-7)) system are differentially expressed during follicular development and can be involved in the follicular health/atresia transition in bovine. The largest (F1) and second largest follicles (F2) were collected from cows before (Day 2), during (Day 3), or after (Day 4) the expected moment of follicular deviation. In the second experiment, F1 was induced to atresia through intrafollicular injection of fulvestrant (estrogen receptor-antagonist) and, in both experiments, mRNA expression of the Mas receptor, ACE2, NEP, and PEP was evaluated in the granulosa and theca cells. The mRNA expression of Mas receptor was upregulated in the granulosa cells of F2 after the establishment of follicular deviation, while PEP mRNA increased during and after the deviation process. The mRNA expression of ACE2 was upregulated in the granulosa cells of F1 during and after the follicular deviation. The mRNA expression of NEP was not regulated in F1 and F2. Mas receptor expression increased in the F1 induced to atresia. mRNA for Mas receptor, ACE2, and PEP are differentially expressed in granulosa cells throughout follicular development and the Mas receptor can be involved with the establishment of follicular dominance.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
β-Estradiol 3-benzoate, ≥97%
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, meets USP testing specifications