  • Search Results
  • Puerarin improves cardiac function through regulation of energy metabolism in Streptozotocin-Nicotinamide induced diabetic mice after myocardial infarction.
Puerarin improves cardiac function through regulation of energy metabolism in Streptozotocin-Nicotinamide induced diabetic mice after myocardial infarction.
Biochemical and biophysical research communications (2015-06-17)
Weili Cheng, Peng Wu, Yingqiang Du, Yunle Wang, Ningtian Zhou, Yingbin Ge, Zhijian Yang
ABSTRACT
It is well recognized that the incidence of heart failure and the risk of death is high in diabetic patients after myocardial infarction (MI). Accumulating evidence showed that puerarin (PUE) has protecting function on both cardiovascular disease and diabetes. The aim of this study is to explore whether puerarin could improve cardiac function in diabetic mice after MI and the underlying mechanism. The left anterior of Streptozotocin (STZ)-Nicotinamide (NA) induced diabetic mice were ligated permanently except for the Shame group. Then the operated mice were randomly treated with PUE or saline. Cardiac function was evaluated by echocardiograph before and at 1, 2, 4 weeks after MI. GLUT4/CD36/p-Akt/PPAR α of the heart was examined after treatment for 4 weeks. The results indicated that PUE significantly increased survival rate, improved cardiac function compared with MI group. Moreover, PUE increased expression and translocation of GLUT4 while attenuated expression and translocation of CD36. Western blot analysis showed that PUE enhanced phosphorylation of Akt and decreased PPAR α. This study demonstrated that PUE improved cardiac function after MI in diabetic mice through regulation of energy metabolism, the possible mechanism responsible for the effect of PUE was increasing the expression and translocation of GLUT4 while attenuating the expression and translocation of CD36.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C