  • Search Results
  • Pharmacological evaluation of methanolic leaf extract of Swietenia mahagoni on acrylamide-induced neuropathic pain in rats.
Pharmacological evaluation of methanolic leaf extract of Swietenia mahagoni on acrylamide-induced neuropathic pain in rats.
Toxicology and industrial health (2013-05-31)
Subburaj Vanitha, Venkata Rathina Kumar Thiagarajan, Arunachalam Muthuraman, Shanmugarajan Krishnan, Ajithadas Aruna, R Tharabai
ABSTRACT
The present study was designed to investigate the antinociceptive effects of methanolic leaf extract of Swietenia mahagoni (MESM) on acrylamide-induced painful neuropathy in rats. The intraperitoneal administration of acrylamide (30 mg/kg; for 24 consecutive days) has been employed for the induction of painful neuropathy. Acrylamide induced nociceptive pain sensitive changes, which have been assessed by hot plate, Von Frey Hair, and tail immersion tests at different time intervals, that is, 0, 6, 12, 18, and 24th day. Furthermore, the biochemical changes, that is, thiobarbituric acid-reactive substances, reduced glutathione, and total calcium levels have been estimated in sciatic nerve tissue on 24th day and histopathological changes have been observed in sciatic nerve tissue sample. MESM and pregabalin have been administered for 14 consecutive days before 1 h of the each acrylamide injection. Administration of acrylamide resulted in significant changes in behavioral and biochemical parameters. Pretreatment of MESM ameliorated acrylamide-induced behavioral, biochemical, and histopathological changes in a dose-dependent manner, which is similar to that of pregabalin-pretreated group. These findings suggested that the neuroprotective effect of S. mahagoni may be due to its potential of antioxidative, calcium channel modulatory, and neuroprotective action.
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, natural, 97%, FG