  • Search Results
  • THREE-YEAR RESULTS OF POLYPOIDAL CHOROIDAL VASCULOPATHY TREATED WITH PHOTODYNAMIC THERAPY: Retrospective Study and Systematic Review.
THREE-YEAR RESULTS OF POLYPOIDAL CHOROIDAL VASCULOPATHY TREATED WITH PHOTODYNAMIC THERAPY: Retrospective Study and Systematic Review.
Retina (Philadelphia, Pa.) (2015-02-27)
Chee Wai Wong, Chui Ming Gemmy Cheung, Ranjana Mathur, Xiang Li, Choi Mun Chan, Ian Yeo, Edmund Wong, Shu Yen Lee, Doric Wong, Tien Yin Wong
ABSTRACT
To evaluate the 3-year outcome in eyes with polypoidal choroidal vasculopathy (PCV) treated with photodynamic therapy with verteporfin. Retrospective study and review of the literature. We performed a retrospective study of patients with PCV who were treated with photodynamic therapy between January 2007 and December 2008. Patients were excluded if they had received photodynamic therapy before the study period, but those who received previous treatment with other modalities (thermal laser or intravitreal therapies) were allowed. The main outcome measures were best-corrected visual acuity, repeat photodynamic therapy, and recurrence of PCV at the end of Years 1, 2, and 3. We further conducted a systematic review of the literature using the terms "polypoidal choroidal vasculopathy" and "photodynamic therapy" and compared the visual outcome of studies over 3 years using meta-analytical methods. The retrospective study included 68 eyes. The mean best-corrected visual acuity was 0.73 ± 0.56 logMAR (20/107, Snellen equivalent) at baseline, 0.73 ± 0.70 logMAR (20/107, Snellen equivalent) at 1 year, 0.96 ± 0.76 logMAR (20/182, Snellen equivalent) at 2 years, and 1.07 ± 0.81 logMAR (20/235, Snellen equivalent) at 3 years. The cumulative recurrence rates of PCV were 16.1% (1 year), 34.9% (2 years), and 52.7% (3 years) and eyes with recurrence were more likely to suffer ≥3 lines loss compared with eyes without recurrence (63.2 vs. 17.6%, P = 0.006). The systematic review summarized results from 48 published studies and our retrospective study. The pooled analysis from 29 studies (316 eyes reporting the 3-year visual outcome) reported mean best-corrected visual acuity improvement of 0.115 logMAR at 1 year (n = 1,669), 0.066 logMAR at 2 years (n = 701), and 0.027 logMAR at 3 years (n = 316). Reported recurrence rates were 5.9% to 50.0% after 1 year, 9.1% to 83.3% after 2 years, and 40.0% to 78.6% after 3 years or longer of follow-up. The visual outcome in eyes with PCV was stable until 2 years, but the outcome at 3 years worsened, particularly in eyes that experienced recurrence.
Sigma-Aldrich
Verteporfin, ≥94% (HPLC)