  • Search Results
  • Antioxidative phytochemicals from Rhododendron oldhamii Maxim. leaf extracts reduce serum uric acid levels in potassium oxonate-induced hyperuricemic mice.
Antioxidative phytochemicals from Rhododendron oldhamii Maxim. leaf extracts reduce serum uric acid levels in potassium oxonate-induced hyperuricemic mice.
BMC complementary and alternative medicine (2015-12-03)
Yu-Tang Tung, Lei-Chen Lin, Ya-Ling Liu, Shang-Tse Ho, Chi-Yang Lin, Hsiao-Li Chuang, Chien-Chao Chiu, Chi-Chang Huang, Jyh-Horng Wu
ABSTRACT
Some of the genus Rhododendron was used in traditional medicine for arthritis, acute and chronic bronchitis, asthma, pain, inflammation, rheumatism, hypertension and metabolic diseases and many species of the genus Rhododendron contain a large number of phenolic compounds and antioxidant properties that could be developed into pharmaceutical products. In this study, the antioxidative phytochemicals of Rhododendron oldhamii Maxim. leaves were detected by an online HPLC-DPPH method. In addition, the anti-hyperuricemic effect of the active phytochemicals from R. oldhamii leaf extracts was investigated using potassium oxonate (PO)-induced acute hyperuricemia. Six phytochemicals, including (2R, 3R)-epicatechin (1), (2R, 3R)-taxifolin (2), (2R, 3R)-astilbin (3), hyposide (4), guaijaverin (5), and quercitrin (6), were isolated using the developed screening method. Of these, compounds 3, 4, 5, and 6 were found to be major bioactive phytochemicals, and their contents were determined to be 130.8 ± 10.9, 105.5 ± 8.5, 104.1 ± 4.7, and 108.6 ± 4.0 mg per gram of EtOAc fraction, respectively. In addition, the four major bioactive phytochemicals at the same dosage (100 mmol/kg) were administered to the abdominal cavity of potassium oxonate (PO)-induced hyperuricemic mice, and the serum uric acid level was measured after 3 h of administration. H&E staining showed that PO-induced kidney injury caused renal tubular epithelium nuclear condensation in the cortex areas or the appearance of numerous hyaline casts in the medulla areas; treatment with 100 mmol/kg of EtOAc fraction, (2R, 3R)-astilbin, hyposide, guaijaverin, and quercitrin significantly reduced kidney injury. In addition, the serum uric acid level was significantly suppressed by 54.1, 35.1, 56.3, 56.3, and 53.2 %, respectively, by the administrations of 100 mmol/kg EtOAc fraction and the derived major phytochemicals, (2R, 3R)-astilbin, hyposide, guaijaverin, and quercitrin, compared to the PO group. The administration of 10 mg/kg benzbromarone, a well-known uricosuric agent, significantly reduced the serum uric acid level by 45.5 % compared to the PO group. The in vivo decrease in uric acid was consistent with free radical scavenging activity, indicating that the major phytochemicals of R. oldhamii leave extracts and the derived phytochemicals possess potent hypouricemic effects, and they could be potential candidates for new hypouricemic agents.
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Hypoxanthine, ≥99.0%
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Hypoxanthine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Xanthine, ≥99%
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium, cubes (in mineral oil), 99.5% trace metals basis
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Xanthine, ≥99.5% (HPLC), purified by recrystallization
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%