  • Search Results
  • Silica Microcapsules for Long-Term, Robust, and Reliable Room Temperature RNA Preservation.
Silica Microcapsules for Long-Term, Robust, and Reliable Room Temperature RNA Preservation.
Advanced healthcare materials (2015-04-23)
Michela Puddu, Wendelin J Stark, Robert N Grass
ABSTRACT
As a consequence of the latest revolutionary discoveries on its functions, RNA is certainly the hottest topic at the moment, being an exceptional tool in biology as well as in medicine. For the various applications, a proper RNA storage is required to prevent the degradation of this extremely unstable molecule. Here a novel freezing-free RNA storage strategy is presented, based on its encapsulation in silica spheres. The silica microcapsules protect the RNA by providing a water-free environment. In this way RNA can be safely stored for prolonged periods of time at ambient and elevated temperatures, maintaining its original integrity, as proved by gel-electrophoresis, capillary electrophoresis, and real-time reverse transcription-polymerase chain reaction (RT-qPCR). The RNA degradation rate at 65 °C in silica microcapsules is approximately ten times smaller in comparison to dry RNA samples or to samples stored in RNAstable matrix, a commercially available product. Moreover, RNA half-life at 65 °C is nearly identical to that of DNA within the silica microcapsules. Samples intended for use in gene expression are compatible with further analysis (RT-qPCR, Sanger sequencing). The novel storage technology permits to safely handle, store, and transport RNA samples, avoiding the expensive shipments and the problems of space presented by freezing-based strategies.
Sigma-Aldrich
Copper(I) chloride, ≥99.995% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Tetraethyl orthosilicate, 99.999% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Sodium bromide, ≥99.99% trace metals basis
Sigma-Aldrich
Tetraethyl orthosilicate, reagent grade, 98%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium bromide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Sodium bromide, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Tetraethyl orthosilicate, ≥99.0% (GC)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Sodium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Copper(I) chloride, AnhydroBeads, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Sodium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Tetraethyl orthosilicate, packaged for use in deposition systems
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%