  • Search Results
  • An indigenous single-vial kit formulation of human serum albumin nanocolloid for use in sentinel lymph node detection.
An indigenous single-vial kit formulation of human serum albumin nanocolloid for use in sentinel lymph node detection.
Nuclear medicine communications (2015-05-02)
Suresh Subramanian, Usha Pandey, Sneha Shah, Venkatesh Rangarajan, Grace Samuel
ABSTRACT
In-situ sentinel lymph node (SLN) detection is an important component in staging cancers of various origins. At present, technetium-99m (Tc)-labeled nanoparticle formulations like sulfur colloid and human serum albumin (HSA) nanocolloid are used in the clinic as SLN tracers. In India, HSA nanocolloid cold kits have so far been imported. This study aims to develop and evaluate an indigenous alternative to imported HSA nanocolloid cold kits for SLN detection/imaging. Production of cold kits was standardized and the product was characterized for its suitability in terms of particle size. Tc-labeling of an in-house HSA nanocolloid was optimized, and the yield and stability of the product were assessed. Animal studies were performed in Wistar rats using the footpad model. Clinical evaluation was performed in 54 patients using a combination of scintigraphic imaging and a hand-held gamma probe. With the optimized protocol, HSA nanocolloids with particle sizes ranging from 50 to 200 nm were obtained. Greater than 90% Tc-labeling yield was obtained in 15 min reactions, and the radiopharmaceutical was stable for up to 24 h after preparation. The animal studies showed similar SLN uptake and improved retention pattern compared with those of the imported Nanocoll radiopharmaceutical. Clinical studies showed detectable 'hot' nodes in 53 of 54 patients, demonstrating sensitivity of the product for clinical utility. In conclusion, this indigenous HSA nanocolloid cold kit is proposed as a logistically favorable alternative to imported kits for SLN detection in the Indian clinical scenario.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Tin(II) chloride dihydrate, ≥99.995% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Tin(II) chloride dihydrate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis