  • Search Results
  • Assessing the bioavailability of polyphenols and antioxidant properties of extra virgin argan oil by simulated digestion and Caco-2 cell assays. Comparative study with extra virgin olive oil.
Assessing the bioavailability of polyphenols and antioxidant properties of extra virgin argan oil by simulated digestion and Caco-2 cell assays. Comparative study with extra virgin olive oil.
Food chemistry (2015-06-05)
Isabel Seiquer, Ascensión Rueda, Manuel Olalla, Carmen Cabrera-Vique
ABSTRACT
Argan oil is becoming increasingly popular in the edible-oil market as a luxury food with healthy properties. This paper analyzes (i) the bioavailability of the polyphenol content and antioxidant properties of extra virgin argan oil (EVA) by the combination of in vitro digestion and absorption across Caco-2 cells and (ii) the protective role of the oil bioaccessible fraction (BF) against induced oxidative stress. Results were compared with those obtained with extra virgin olive oil (EVO). Higher values of polyphenols and antioxidant activity were observed in the BF obtained after the in vitro digestion of oils compared with the initial chemical extracts; the increase was higher for EVA but absolute BF values were lower than EVO. Bioaccessible polyphenols from EVA were absorbed by Caco-2 cells in higher proportions than from EVO, and minor differences were observed for antioxidant activity. Preincubation of cell cultures with BF from both oils significantly protected against oxidation, limiting cell damage and reducing reactive oxygen species generation.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Luperox® TBH70X, tert-Butyl hydroperoxide solution, 70 wt. % in H2O
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in decane
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
SAFC
HEPES
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, for spectrophotometric det. (of Fe), ≥98%
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
tert-Butyl hydroperoxide solution, packed in FEP bottles, ~5.5 M in decane (over molecular sieve 4Å)
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
SAFC
HEPES
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Sigma-Aldrich
Sodium bicarbonate, Hybri-Max, powder, suitable for hybridoma, ≥99.5%
Sigma-Aldrich
CACO-2, Caucasian colon adenocarcinoma
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in nonane
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%