  • Search Results
  • A carbon nanotube/poly [Ni-(Protoporphyrin IX)] composite for amperometric detection of long chain aliphatic amines.
A carbon nanotube/poly [Ni-(Protoporphyrin IX)] composite for amperometric detection of long chain aliphatic amines.
Bioelectrochemistry (Amsterdam, Netherlands) (2015-04-02)
Romina Carballo, Ana L Rinaldi, Paula C Dabas, Irene N Rezzano
ABSTRACT
Poly [Ni-Protoporphyrin] film (pNiPP), containing multiwall carbon nanotubes (MWCNT) was used to cover a glassy carbon electrode. The hybrid material (pNiPP/MWCNT) successfully combines the permselectivity of pNiPP with the high conductivity of MWCNT. The modified electrode was used to perform amperometric detection of long chain aliphatic amines (LCAA) in order to prevent the passivation effect of the aliphatic chain. Cyclic voltammetry (CV) and electrochemical impedance spectroscopy (EIS) demonstrated that the pNiPP/MWCNT facilitates the electron transfer reaction. The charge transfer resistance (Rct) values were significantly lower by up to one order of magnitude compared to the bare electrode. Differential pulse polarography (DPP) showed a marked decrease of the overpotential generated by the aliphatic chain. The calibration of the amperometric peak area vs. concentrations of derivatized LCAA exhibits a linear response within the range of 0.018 and 28 μM and correlation coefficient (R(2)) higher than 0.999 (n=5). The quantitation limit of the pNiPP/MWCNT electrode is about 400 times lower than the UV-visible detection. RSD of 7.2%, 5.8%, 2.5% and 2.3% was obtained for concentrations of 0.028, 0.28, 2.8 and 28 μM of ferrocenyl octadecylamine. A solution of sphingosine, 0.23 μM, was exclusively detected with HPLC-ECD with pNiPP/MWCNT electrode.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Hydroxylamine solution, 50 wt. % in H2O
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Lithium perchlorate, battery grade, dry, 99.99% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Octadecylamine, ≥99% (GC)
Sigma-Aldrich
Hydroxylamine solution, 50 wt. % in H2O, 99.999%
Sigma-Aldrich
Hexadecylamine, 98%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Ferrocenemethanol, 97%
Sigma-Aldrich
Lithium perchlorate, 99.99% trace metals basis
Sigma-Aldrich
Octadecylamine, ≥99.0% (GC)
Sigma-Aldrich
Ferrocenecarboxaldehyde, 98%
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Octadecylamine, technical grade, 90%
Sigma-Aldrich
Hexadecylamine, technical grade, 90%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Sodium borohydride, hydrogen-storage grade, 98%
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
VenPure® SF, powder
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.