  • Search Results
  • Down-regulation of Rab17 promotes tumourigenic properties of hepatocellular carcinoma cells via Erk pathway.
Down-regulation of Rab17 promotes tumourigenic properties of hepatocellular carcinoma cells via Erk pathway.
International journal of clinical and experimental pathology (2015-07-21)
Jingsong Qi, Peng Zhao, Fenbao Li, Yingchang Guo, Hongkai Cui, Aiguang Liu, Huajie Mao, Yongli Zhao, Xizhong Zhang
ABSTRACT
The small GTPase, Ras-related protein 17 (Rab17), a member of the Rab family, plays a critical role in the regulation of membrane traffic in polarized eukaryotic cells. However, the role of Rab17 in hepatocellular carcinoma (HCC) is not clear. Clinical speciments reveal that Rab17 was present in 15 of 20 (75.0%) paraneoplastic tissues and 7 of 20 (35.0%) HCC samples (P=0.0248). To elucidate the tumourigenic role of Rab17 in HCC, we generated two Rab17 low-expressing HCC cell lines (Hep3B and Huh-7). The results showed that Rab17 down-regulation significantly promoted the tumourigenic properties of HCC cells in vitro and in vivo, as demonstrated by enhanced cell proliferation, colony formation, invasion and migration, decreased G1 arrest, and increased tumour xenograft growth and angiogenesis. However, the enhanced tumourigenic properties of HCC cells by Rab17 down-regulation was significantly inhibited by PD980592, the inhibitor of the Erk pathway, indicating that the Erk pathway plays a critical role in Rab17 down-regulation-induced enhanced tumourigenic properties of HCC cells. Our data provide a new insight into the essential role of Rab17 in HCC carcinogenesis and suggest that Rab17 expression might be tumor suppressor gene and might provide a new interventional therapeutic target for this common malignancy.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)