Menstrual cyclic change of metastin/GPR54 in endometrium.
Medical molecular morphology (2014-06-09)
Tsukasa Baba, Hyun Sook Kang, Yuko Hosoe, Budiman Kharma, Kaoru Abiko, Noriomi Matsumura, Junzo Hamanishi, Ken Yamaguchi, Yumiko Yoshioka, Masafumi Koshiyama, Masaki Mandai, Susan K Murphy, Ikuo Konishi
ABSTRACT
Metastin/kisspeptin is encoded by KISS1 and functions as an endogenous ligand of GPR54. Interaction of metastin with GPR54 suppresses metastasis and also regulates release of gonadotropin-releasing hormone, which promotes secretion of estradiol (E2) and progesterone (P4). We have previously demonstrated epigenetic regulation of GPR54 in endometrial cancer and the potent role of metastin peptides in inhibiting metastasis in endometrial cancer. However, little is known about how the metastin-GPR54 axis is regulated in the endometrium, the precursor tissue of endometrial cancer. Endometrial stromal cells (ESCs) and endometrial glandular cells (EGCs) within the endometrium show morphological changes when exposed to E2 and P4. In this study, we show that metastin expression is induced in ESCs through decidualization, but is repressed in glandular components of atypical endometrial hyperplasia (AEH) and endometrial cancer relative to EGCs. The promoter of GPR54 is unmethylated in normal endometrium and in AEH. These results indicate metastin may function in decidualized endometrium to prepare for adequate placentation but this autocrine secretion of metastin is deregulated during oncogenesis to enable tumor cells to spread.
Sigma-Aldrich
Propylene glycol monomethyl ether acetate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Medroxyprogesterone 17-acetate, ≥97% (HPLC)
Sigma-Aldrich
Phenol, natural, 97%, FG