Cortisol Measures Across the Weight Spectrum.
The Journal of clinical endocrinology and metabolism (2015-07-15)
Melanie Schorr, Elizabeth A Lawson, Laura E Dichtel, Anne Klibanski, Karen K Miller
ABSTRACT
There are conflicting reports of increased vs decreased hypothalamic-pituitary-adrenal (HPA) activation in obesity; the most consistent finding is an inverse relationship between body mass index (BMI) and morning cortisol. In anorexia nervosa (AN), a low-BMI state, cortisol measures are elevated. This study aimed to investigate cortisol measures across the weight spectrum. This was a cross-sectional study at a clinical research center. This study included 60 women, 18-45 years of age: overweight/obese (OB; N = 21); AN (N = 18); and normal-weight controls (HC; N = 21). HPA dynamics were assessed by urinary free cortisol, mean overnight serum cortisol obtained by pooled frequent sampling every 20 minutes from 2000-0800 h, 0800 h serum cortisol and cortisol-binding globulin, morning and late-night salivary cortisol, and dexamethasone-CRH testing. Body composition and bone mineral density (BMD) were assessed by dual-energy x-ray absorptiometry. Cortisol measures demonstrated a U-shaped relationship with BMI, nadiring in the overweight-class I obese range, and were similarly associated with visceral adipose tissue and total fat mass. Mean cortisol levels were higher in AN than OB. There were weak negative linear relationships between lean mass and some cortisol measures. Most cortisol measures were negatively associated with postero-anterior spine and total hip BMD. Cortisol measures are lowest in overweight-class I obese women-lower than in lean women. With more significant obesity, cortisol levels increase, although not to as high as in AN. Therefore, extreme underweight and overweight states may activate the HPA axis, and hypercortisolemia may contribute to increased adiposity in the setting of caloric excess. Hypercortisolemia may also contribute to decreased BMD and muscle wasting in the setting of both caloric restriction and excess.
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications