  • Search Results
  • Effect of R-CHOP chemotherapy on liver and mediastinal blood pool (18)F-FDG standardized uptake values in patients with non-Hodgkin's lymphoma.
Effect of R-CHOP chemotherapy on liver and mediastinal blood pool (18)F-FDG standardized uptake values in patients with non-Hodgkin's lymphoma.
Clinical imaging (2015-02-25)
Bugra Kaya, Zeki Dostbil, Murat Ismailoglu, Bekir Tasdemir, Ozlem Sahin
ABSTRACT
We aimed to investigate the impact of chemotherapy on (18)F-FDG uptake in the liver and mediastinal blood pool (MBP) among patients with non-Hodgkin's lymphoma. Twenty-three patients with NHL underwent baseline, interim, and postchemotherapy (18)F-FDG PET/CT. SUVmax and SUVmean values of the liver and MBP at imaging time were compared statistically. We did not find any significant differences between the liver and mediastinum SUVmean and SUVmax values (P>.05). Our study demonstrates that the (18)F-FDG uptake in the liver and MBP are not significantly affected by R-CHOP chemotherapy in patients with NHL.
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Tantalum(V) ethoxide, packaged for use in deposition systems