  • Search Results
  • Conceptualizing the neurobiology of non-suicidal self-injury from the perspective of the Research Domain Criteria Project.
Conceptualizing the neurobiology of non-suicidal self-injury from the perspective of the Research Domain Criteria Project.
Neuroscience and biobehavioral reviews (2015-10-02)
Melinda Westlund Schreiner, Bonnie Klimes-Dougan, Erin D Begnel, Kathryn R Cullen
ABSTRACT
Non-suicidal self-injury (NSSI) commonly starts in adolescence and is associated with an array of negative outcomes. Neurobiological research investigating NSSI is in its early stages and most studies have examined this behavior within the context of specific diagnoses. However, the Research Domain Criteria (RDoC) initiative encourages researchers to examine brain-behavior relationships across diagnoses. This review on the neurobiology associated with NSSI is organized using the domains proposed by RDoC: Negative Valence, Positive Valence, Cognitive, Social Processes, and Arousal/Regulatory Systems. Evidence of neurobiological anomalies is found in each of these domains. We also propose future research directions, especially in regard to human development. Future NSSI studies should address this behavior independent of diagnosis, examine relevant constructs across multiple units of analysis, and assess how systems change across development and course of illness. These advances will be essential for guiding neurobiologically informed intervention and prevention strategies to target NSSI. In doing so, we may prevent the associated negative outcomes across the lifespan.
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications