  • Search Results
  • Aberrant CD200/CD200R1 expression and its potential role in Th17 cell differentiation, chemotaxis and osteoclastogenesis in rheumatoid arthritis.
Aberrant CD200/CD200R1 expression and its potential role in Th17 cell differentiation, chemotaxis and osteoclastogenesis in rheumatoid arthritis.
Rheumatology (Oxford, England) (2014-09-28)
Yan Ren, Bo Yang, Yufeng Yin, Xiaomei Leng, Ying Jiang, Lei Zhang, Yongzhe Li, Xin Li, Fengchun Zhang, Wei He, Xuan Zhang, Xuetao Cao
ABSTRACT
CD200/CD200R1 signalling has an immunoregulatory effect on the activation threshold of the inflammatory immune response and maintains immune homeostasis. In this study we evaluated the status of CD200/CD200R1 interaction in patients with RA. The expression of CD200 and CD200R1 was examined by immunohistochemistry and flow cytometry and was compared between RA patients and healthy controls (HCs). Sorted CD4(+) T cells were stained with carboxyfluorescein succinimidyl ester (CFSE) and annexin V-propidium iodide to evaluate the effect of CD200 on cell proliferation and apoptosis. The effect of CD200 on Th17 differentiation, function and osteoclastogenesis was determined by flow cytometry, transwell migration assay and immunocytochemistry, respectively. The proportion of CD200(+) cells and CD200R1(+) cells in peripheral blood mononuclear cells, peripheral CD14(+) cells and CD4(+) T cells was significantly lower in the RA patients than in HCs, whereas the number of CD200(+) cells was higher in synovium from RA patients than in that from HCs. After treatment with infliximab and MTX we found increased expression of peripheral CD200/CD200R1 that correlated with a decrease in the 28-joint DAS. CD200Fc in vitro partially inhibited CD4(+) T cell proliferation, promoted CD4(+) T cell apoptosis, reduced CD4(+) T cell differentiation into Th17 cells and down-regulated CCR6-mediated Th17 chemotaxis in cells from RA patients. In addition, the engagement of the CD200 receptors on CD14(+) cells with CD200Fc in vitro reduced osteoclastogenesis and inhibited CD14(+) cell-driven Th17 differentiation. Abnormal CD200/CD200R1 expression in RA may contribute to abnormal Th17 cell differentiation, chemotaxis and osteoclastogenesis.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
IL-2 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in Pichia pastoris, suitable for cell culture
Sigma-Aldrich
Interleukin-2 from mouse, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in HEK 293 cells, suitable for cell culture, endotoxin tested
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in E. coli, ~10000 U/mL
Sigma-Aldrich
IL-2 from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
Interleukin-2 from mouse, IL-2, recombinant, expressed in E. coli, carrier free
Sigma-Aldrich
IL-2 human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)