  • Search Results
  • Clusterin/apolipoprotein J binds to aggregated LDL in human plasma and plays a protective role against LDL aggregation.
Clusterin/apolipoprotein J binds to aggregated LDL in human plasma and plays a protective role against LDL aggregation.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2015-01-01)
Maria Martínez-Bujidos, Anna Rull, Beatriz González-Cura, Montserrat Pérez-Cuéllar, Laia Montoliu-Gaya, Sandra Villegas, Jordi Ordóñez-Llanos, José Luis Sánchez-Quesada
ABSTRACT
Clusterin/apolipoprotein J (apoJ) is an extracellular chaperone involved in the quality control system against protein aggregation. A minor part of apoJ is transported in blood bound to LDLs, but its function is unknown. Our aim was to determine the role of apoJ bound to LDLs. Total LDL from human plasma was fractionated into native LDL [LDL(+)] and electronegative LDL [LDL(-)]. The latter was separated into nonaggregated [nagLDL(-)] and aggregated LDL(-) [agLDL(-)]. The content of apoJ was 6-fold higher in LDL(-) than in LDL(+) and 7-fold higher in agLDL(-) than in nagLDL(-). The proportion of LDL particles containing apoJ (LDL/J+) was 3-fold lower in LDL(+) than in LDL(-). LDL/J+ particles shared several characteristics with agLDL(-), including increased negative charge and aggregation. apoJ-depleted particles (LDL/J-) showed increased susceptibility to aggregation, whether spontaneous or induced by proteolysis or lipolysis, as was revealed by turbidimetric analysis, gel filtration chromatography, lipoprotein precipitation, native gradient gel electrophoresis, circular dichroism, and transmission electronic microscopy. The addition of purified apoJ to total LDL also prevented its aggregation induced by proteolysis or lipolysis. These findings point to apoJ as a key modulator of LDL aggregation and reveal a putative new therapeutic strategy against atherosclerosis.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, ≥99.0% (GC), powder
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
SAFC
Cholesterol, Plant-Derived, SyntheChol®, PharmaGrade, USP/NF, Ph. Eur., Manufactured under appropriate GMP controls for pharma or biopharmaceutical production
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
SAFC
Glycine, PharmaGrade, Ajinomoto, EP, JP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.