  • Search Results
  • NKP30-B7-H6 Interaction Aggravates Hepatocyte Damage through Up-Regulation of Interleukin-32 Expression in Hepatitis B Virus-Related Acute-On-Chronic Liver Failure.
NKP30-B7-H6 Interaction Aggravates Hepatocyte Damage through Up-Regulation of Interleukin-32 Expression in Hepatitis B Virus-Related Acute-On-Chronic Liver Failure.
PloS one (2015-08-05)
Yong Zou, Junjie Bao, Xingfei Pan, Ying Lu, Sihong Liao, Xicheng Wang, Guoying Wang, Dongjun Lin
ABSTRACT
Previous work conducted by our group has shown that the accumulation of hepatic natural killer (NK) cells and the up-regulation of natural cytotoxicity receptors (NKP30 and NKP46) on NK cells from patients with hepatitis B virus-related acute-on-chronic liver failure (HBV-ACLF) were correlated with disease progression in HBV-ACLF. The natural cytotoxicity receptors expressed on NK cells are believed to be probable candidates involved in the NK cell-mediated hepatocyte damage in HBV-ACLF. However, the underlying mechanisms remain to be elucidated. In the present study, we aimed to discover the role of NKP30-B7-H6 interaction in NK cells-mediated hepatocyte damage in HBV-ACLF. Hepatic expressions of B7-H6 and interleukin-32 (IL-32) were examined by immunochemistry staining in samples from patients with HBV-ACLF or mild chronic hepatitis B (CHB). The cytotoxicity of NK-92 cell against target cells (Huh-7 and LO2) was evaluated by CCK8 assay. Expression of IL-32 in liver NK cell, T cells and NK-92 cell line was detected by the flow cytometric analysis. The effect of IL-32 on the apoptosis of Huh7 cells was evaluated using Annexin V/PI staining analysis. An enhancement of hepatic B7-H6 and IL-32 expression was associated with the severity of liver injury in HBV-ACLF. And there was a positive association between hepatic B7-H6 and IL-32 expression. Expressions of IL-32 in liver NK cells and T cells were increased in HBV-ACLF patients. In vitro NK-92 cells are highly capable of killing the high B7-H6 expressing Huh7 cells and B7-H6-tansfected hepatocyte line LO2 cells dependent on NKP30 and B7-H6 interaction. Furthermore, NK-92 cells exhibited elevated IL-32 expression when stimulated with anti-NKP30 antibodies or when co-cultured with Huh7 cells. IL-32 can induce the apoptosis of Huh7 cells in a dose-dependent manner. Our results suggest that NKP30-B7-H6 interaction can aggravate hepatocyte damage, probably through up-regulation of IL-32 expression in HBV-ACLF.
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
IL-2 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in Pichia pastoris, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in HEK 293 cells, suitable for cell culture, endotoxin tested
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in E. coli, ~10000 U/mL
Sigma-Aldrich
IL-2 human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)