  • Search Results
  • Biobased carbon content of resin extracted from polyethylene composite by carbon-14 concentration measurements using accelerator mass spectrometry.
Biobased carbon content of resin extracted from polyethylene composite by carbon-14 concentration measurements using accelerator mass spectrometry.
SpringerPlus (2015-02-13)
Kazuhiro Taguchi, Masao Kunioka, Masahiro Funabashi, Fumi Ninomiya
ABSTRACT
An estimation procedure for biobased carbon content of polyethylene composite was studied using carbon-14 ((14)C) concentration ratios as measured by accelerated mass spectrometry (AMS). Prior to the measurement, additives and fillers in composites should be removed because they often contain a large amount of biobased carbon and may shift the estimation. Samples of resin with purity suitable for measurement were isolated from composites with a Soxhlet extractor using heated cyclohexanone. After cooling of extraction solutions, the resin was recovered as a fine semi-crystalline precipitate, which was easily filtered. Recovery rates were almost identical (99%), even for low-density polyethylene and linear low-density polyethylene, which may have lower crystallinity. This procedure could provide a suitable approach for estimation of biobased carbon content by AMS on the basis of the standard ASTM D 6866. The biobased carbon content for resin extracted from polyethylene composites allow for the calculation of biosynthetic polymer content, which is an indicator of mass percentage of the biobased plastic resin in the composite.
Sigma-Aldrich
Calcium carbonate, ACS reagent, chelometric standard, 99.95-100.05% dry basis
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, >40 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
1-Butene, ≥99%
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 200 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), beads
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 35 μm particle size
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, ≥350 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), ≤12 μm particle size