  • Search Results
  • An integrative approach identified genes associated with drug response in gastric cancer.
An integrative approach identified genes associated with drug response in gastric cancer.
Carcinogenesis (2015-03-07)
Jin Zhou, Wei-Peng Yong, Chui Sun Yap, Aadhitthya Vijayaraghavan, Rohit Anthony Sinha, Brijesh Kumar Singh, Sam Xiu, Sravanthy Manesh, Anna Ngo, Andrea Lim, Carolyn Ang, Chen Xie, Foong Ying Wong, Suling J Lin, Wei Keat Wan, Iain Beehuat Tan, Horst Flotow, Patrick Tan, Kiat-Hon Lim, Paul Michael Yen, Liang Kee Goh
ABSTRACT
Gastric cancer (GC) is the second leading cause of global cancer mortality worldwide. However, the molecular mechanism underlying its carcinogenesis and drug resistance is not well understood. To identify novel functionally important genes that were differentially expressed due to combinations of genetic and epigenetic changes, we analyzed datasets containing genome-wide mRNA expression, DNA copy number alterations and DNA methylation status from 154 primary GC samples and 47 matched non-neoplastic mucosa tissues from Asian patients. We used concepts of 'within' and 'between' statistical analysis to compare the difference between tumors and controls within each platform, and assessed the correlations between platforms. This 'multi-regulated gene (MRG)' analysis identified 126 differentially expressed genes that underwent a combination of copy number and DNA methylation changes. Most genes were located at genomic loci associated with GC. Statistical enrichment analysis showed that MRGs were enriched for cancer, GC and drug response. We analysed several MRGs that previously had not been associated with GC. Knockdown of DDX27, TH1L or IDH3G sensitized cells to epirubicin or cisplatin, and knockdown of RAI14 reduced cell proliferation. Further studies showed that overexpression of DDX27 reduced epirubicin-induced DNA damage and apoptosis. Levels of DDX27 mRNA and protein were increased in early-stage gastric tumors, and may be a potential diagnostic and prognostic marker for GC. In summary, we used an integrative bioinformatics strategy to identify novel genes that are altered in GC and regulate resistance of GC cells to drugs in vitro.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Trimesic acid, 95%
Sigma-Aldrich
Trimethylaluminum, 97%
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in toluene
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in hexanes
Sigma-Aldrich
Trimethylaluminum, packaged for use in deposition systems
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in heptane