  • Search Results
  • Marker-ion analysis for quantification of mucoadhesivity of microparticles in particle-retention assays.
Marker-ion analysis for quantification of mucoadhesivity of microparticles in particle-retention assays.
International journal of pharmaceutics (2015-04-18)
Daniel Preisig, Michael Weingartner, Felipe J O Varum, Roberto Bravo, Rainer Alles, Jörg Huwyler, Maxim Puchkov
ABSTRACT
The objective of the present work was to develop an improved method to quantify particle retention on mucosal tissue under dynamic flow conditions with simultaneous determination of drug dissolution. The principle was to dissolve the collected inert carrier material and quantify specific marker ions by reliable analytical methods. The mucoadhesive model particles consisted of drug-loaded porous calcium carbonate microcarriers coated with chitosan, and quantification of calcium ions by capillary electrophoresis enabled to determine particle-retention kinetics on colonic mucosal tissue. The method was validated by image analysis, and the particle-retention assay was successfully applied to granulate material (125-250 mm) and small particles (<90 μm) with mucoadhesive properties. Particle retention on colonic mucosa was improved by increasing the chitosan content, demonstrating the sensitivity and usefulness of marker-ion analysis for quantification of detached particles. Furthermore, we showed that drug dissolution from mucoadhesive microparticles followed comparable kinetics in the particle-retention assay and the standard USP IV method. Our findings are helpful for the development of micro-sized colonic drug delivery systems, in particular for optimization of mucoadhesive properties and sustained drug release kinetics of porous drug carriers.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Phthaldialdehyde, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder
Sigma-Aldrich
Phthaldialdehyde, ≥97% (HPLC), powder (may contain lumps)
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Phthaldialdehyde, for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Boric acid, 99.97% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Boric acid, 99.999% trace metals basis
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%