  • Search Results
  • Lack of tissue accumulation of grape seed flavanols after daily long-term administration in healthy and cafeteria-diet obese rats.
Lack of tissue accumulation of grape seed flavanols after daily long-term administration in healthy and cafeteria-diet obese rats.
Journal of agricultural and food chemistry (2015-10-27)
Maria Margalef, Zara Pons, Lisard Iglesias-Carres, Francisca Isabel Bravo, Begoña Muguerza, Anna Arola-Arnal
ABSTRACT
After ingestion flavanols are metabolized by phase-II enzymes and the microbiota and are distributed throughout the body depending on several factors. Herein we aim to evaluate whether flavanols are tissue-accumulated after the long-term administration of a grape seed polyphenol extract (GSPE) in rats and to study if compounds present in tissues differ in a cafeteria-diet obesity state. For that, plasma, liver, mesenteric white adipose tissue (MWAT), brain, and aorta flavanol metabolites from standard chow-diet-fed (ST) and cafeteria-diet-fed (CAF) rats were analyzed by high-performance liquid chromatography electrospray ionization tandem mass spectrometry (HPLC-ESI-MS/MS) 21 h after the last 12-week-daily GSPE (100 mg/kg) dosage. Results showed that long-term GSPE intake did not trigger a flavanol tissue accumulation, indicating a clearance of products at each daily dosage. Therefore, results suggest that polyphenol benefits in a disease state would be due to a daily pulsatile effect. Moreover, obesity induced by diet also influences the metabolism and bioavailability of flavanols in rats.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
Sigma-Aldrich
Propionic acid, ≥99.5%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%