  • Search Results
  • Lentivirus-mediated gene silencing of NOB1 suppresses non-small cell lung cancer cell proliferation.
Lentivirus-mediated gene silencing of NOB1 suppresses non-small cell lung cancer cell proliferation.
Oncology reports (2015-07-17)
Weiyi Huang, Weiqing Zhong, Jun Xu, Benhua Su, Guanghui Huang, Jiajun Du, Qi Liu
ABSTRACT
NIN/RPN12 binding protein 1 (NOB1p) encoded by NOB1 has been found to be an essential factor in 26S proteasome biogenesis which participates in protein degradation. However, the functions of NOB1 in non-small cell lung cancer cells are largely unknown. In the present study, lentivirus-mediated NOB1 shRNA transfection in two non-small cell lung cancer cell lines (A549 and H1299) was accomplished, as determined by fluorescence imaging. Downregulation of NOB1 expression was confirmed by real-time PCR and western blotting. NOB1 silencing resulted in a significant decline in the proliferation and colony formation capability of non-small cell lung cancer cells. Moreover, flow cytometry showed that A549 cells were arrested in the G0/G1 phase of the cell cycle after NOB1 suppression. Furthermore, depletion of NOB1 resulted in a significant decrease in CDK4 and cyclin D1 expression. These results suggest that NOB1 may act as an important regulator in non-small cell lung cancer growth and could be a therapeutic target of non‑small cell lung cancer.
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Nitrosobenzene, ≥97%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade