  • Search Results
  • Effects of Dietary Supplementation with Brazil Nuts on Microvascular Endothelial Function in Hypertensive and Dyslipidemic Patients: A Randomized Crossover Placebo-Controlled Trial.
Effects of Dietary Supplementation with Brazil Nuts on Microvascular Endothelial Function in Hypertensive and Dyslipidemic Patients: A Randomized Crossover Placebo-Controlled Trial.
Microcirculation (New York, N.Y. : 1994) (2015-07-28)
Grazielle V B Huguenin, Annie S B Moreira, Tatiana D Siant'Pierre, Rodrigo A Gonçalves, Glorimar Rosa, Glaucia M M Oliveira, Ronir R Luiz, Eduardo Tibirica
ABSTRACT
To investigate the effects of dietary supplementation with GBNs on microvascular endothelial function in hypertensive and dyslipidemic patients. Ninety-one patients of both sexes aged 62.1 ± 9.3 years received 13 g/day of GBNs or a placebo for three months with a washout period of one month between treatments. Microvascular endothelial function was assessed using LSCI coupled with iontophoresis of ACh and PORH. We also used skin video capillaroscopy to measure capillary density and recruitment at rest and during PORH. Plasma concentrations of NOx were also measured as a marker of nitric oxide bioavailability. Supplementation with GBNs significantly increased the plasma levels of Se (p < 0.05) and NOx (p < 0.05). However, we did not observe any effects of GBN consumption on microvascular vasodilator responses to ACh or PORH (p > 0.05), and GBNs did not improve capillary density at baseline or recruitment during PORH (p > 0.05). Supplementation with GBNs induced significant increases in the plasma Se concentration and systemic bioavailability of nitric oxide. Nevertheless, GBN supplementation did not lead to any improvement in systemic microvascular reactivity or density in patients with arterial hypertension and dyslipidemia who were undergoing multiple drug therapies.
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, red, fuming, HNO3 >90 %
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., for determinations with dithizone, ≥65%
Sigma-Aldrich
Nitric acid, puriss. p.a., ≥65% (T)
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Acetylcholine bromide, ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N