  • Search Results
  • Determination of the active transport of fucoidan derived from Okinawa Mozuku across the human intestinal Caco-2 cells as assessed by size-exclusion chromatography.
Determination of the active transport of fucoidan derived from Okinawa Mozuku across the human intestinal Caco-2 cells as assessed by size-exclusion chromatography.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-06-30)
Takeaki Nagamine, Kou Hayakawa, Kyoumi Nakazato, Masahiko Iha
ABSTRACT
In order to clarify the mechanism of fucoidan transport, we developed the chromatographic determination method. A size-exclusion chromatography (SEC) method for the determination of Okinawa-fucoidan using Develosil 300 Diol-5 (60×8.0mm I.D., 30nm pore-diameter) with the eluent containing 1% non-ionic detergent is developed. Determination range (UV at 210nm) is from 0 to 100ng of fucoidan with the linear calibration line inserting to zero. A transport activity of fucoidan is demonstrated by using Caco-2 cells (model of gut transport system); i.e., the initial transport velocity 12nmol/h/mg of protein (25-fold slower rate as compared to a bacterial l-alanine active-transport activity 300nmol/h/mg of protein) is found to occur. Since this fucoidan transport is inhibited by 10mM sodium azide (respiration inhibitor) and 0.05mM FCCP (uncoupler), this transport by Caco-2 cells is found to be an active one requiring energy-source. On the other hand, colchicine (inhibitor of phagocytosis/pinocytosis) and mannitol (putative competitive-inhibitor of tight-junction transport) cannot inhibit the fucoidan transport at all. We firstly report that the active transport occurs for such a high molecular-weight sulphated-polyfucose of fucoidan in vitro using Caco-2 cells.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%