  • Search Results
  • Differential acute effects of sleep on spontaneous and stimulated production of tumor necrosis factor in men.
Differential acute effects of sleep on spontaneous and stimulated production of tumor necrosis factor in men.
Brain, behavior, and immunity (2014-12-17)
Stoyan Dimitrov, Luciana Besedovsky, Jan Born, Tanja Lange
ABSTRACT
Tumor necrosis factor (TNF) is considered a key molecule in the regulation of sleep in health and disease. Conversely, sleep compared to sleep deprivation can modulate TNF release, but overall results are conflicting. In this study we focused on the influence of sleep on spontaneous, i.e., unstimulated TNF production, which might be involved in sleep regulation under normal non-infectious conditions, and on lipopolysaccharide (LPS)-stimulated TNF production, which reflects the capacity of the immune system to respond to a pathogen. To this end, we monitored 10 healthy men during a regular sleep-wake cycle and during 24h of wakefulness while blood was sampled repeatedly to analyze circulating TNF levels in serum as well as intracellular TNF production in monocytes spontaneously and after stimulation with LPS employing whole blood cell cultures. In addition we assessed numbers of monocyte subsets and levels of various hormones in blood. In comparison with nocturnal wakefulness, sleep acutely decreased serum TNF levels, with no parallel decrease in spontaneous monocytic TNF production, but was associated with a striking nighttime increase in the percentage of TNF producing monocytes after stimulation with LPS. The following day circulating TNF showed a reverse pattern with higher levels after regular sleep than after the nocturnal vigil. The mechanisms mediating the differential effects of sleep on circulating TNF (acutely decreased) vs. stimulated monocytic TNF production (acutely increased) remain unclear, although explorative correlational analyses pointed to a regulatory involvement of cortisol, norepinephrine and prolactin. The acute enhancing effect of sleep on LPS stimulated monocytic TNF production adds to the notion that nocturnal sleep favors immune defense to a microbial challenge.
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO