  • Search Results
  • Steroidogenesis and phase II conjugation during the gametogenesis of thicklip grey mullet (Chelon labrosus) from a population showing intersex condition.
Steroidogenesis and phase II conjugation during the gametogenesis of thicklip grey mullet (Chelon labrosus) from a population showing intersex condition.
General and comparative endocrinology (2015-01-27)
Adriana E Sardi, Cristina Bizarro, Miren P Cajaraville, Maren Ortiz-Zarragoitia
ABSTRACT
Steroidogenesis, the process by which steroid hormones are synthesized, involves a vast number of enzymes and biochemical pathways that are susceptible to chemical modulation. Endocrine disrupting compounds (EDCs) are of special concern since they can alter hormone homeostasis by interfering with synthesis, transport and elimination of hormones. It is important to understand gender differences and the natural variation in steroid balance through gamete development in fish exposed to EDCs. The aim of this study was to determine mRNA levels of genes encoding for Steroidogenic Acute Regulatory (star) protein; the steroidogenic enzymes P450 11β hydroxylase (cyp11b1) and P450 aromatase (cyp19a1a); as well as the phase II conjugation enzymes sulfotransferase (sult) and UDP-glucuronosyltransferase (ugt), together with the activity of P450 aromatase and plasma levels of 17β-estradiol (E2) and 11-ketotestosterone (11-KT), at different gametogenic stages and in intersex individuals of the thicklip grey mullet Chelon labrosus. Results demonstrated that the transcription levels of star, sult and ugt and levels of E2 and 11-KT in plasma significantly changed with the interaction between gender and reproductive stage. Cyp11b1 and cyp19a1a transcription levels were significantly different between genders while the activity of P450 aromatase varied significantly between genders and reproductive stages. Results from a multivariate assessment demonstrated that measured endpoints distinguished male, female and intersex mullets at immature gametogenic stage. Intersex distinction was based on sult, ugt and cyp19a1a transcript levels and P450 aromatase activity. The present work provides data to be used in future experimental designs with C. labrosus species, and gives new clues about the molecular events that lead to intersex occurrence in mullets.
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Zirconium(IV) oxide, powder, 5 μm, 99% trace metals basis
Sigma-Aldrich
Zirconium(IV) oxide, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Zirconium(IV) oxide, 99.99% trace metals basis (purity excludes ~2% HfO2)
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 10 wt. % in H2O
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 5 wt. % in H2O
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Ethyl 3-aminobenzoate methanesulfonate, 98%
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
11-Ketotestosterone
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥98%