  • Search Results
  • Amorphous solid dispersion with increased gastric solubility in tandem with oral disintegrating tablets: a successful approach to improve the bioavailability of atorvastatin.
Amorphous solid dispersion with increased gastric solubility in tandem with oral disintegrating tablets: a successful approach to improve the bioavailability of atorvastatin.
Pharmaceutical development and technology (2014-02-05)
Jumah Masoud M Salmani, Huixia Lv, Sajid Asghar, Jianping Zhou
ABSTRACT
Serious efforts have been made to overcome the bioavailability problems of ever increasing number of poorly soluble drugs, including atorvastatin (ATO); however, enhancing its gastric solubility has not received much attention. To improve the bioavailability of ATO by increasing its gastric solubility in a stable oral disintegration tablet (ODT) formulation. Amorphous solid dispersion (ASD) of ATO with Eudragit® EPO was used as API in ODT formulation. Characterization using Differential scanning calorimetry, Powder X-ray diffraction, Fourier transform infrared drug-polymer interaction simulated by molecular modeling, solubility, dissolution and stability studies together with in vivo evaluation. In ASD there was uniform distribution of drug in the polymer and it retained the amorphous nature without any chemical interactions except the possibility of hydrogen bond formation, with substantially higher gastric solubility. The dissolution profile of the ODT containing ASD was significantly improved >90% within 15 min compared with 25% of plain ATO formulation. In vivo results showed an overall enhancement in the apparent bioavailability (83% and 434% more than Lipitor® and plain amorphous ATO tablets, respectively). Combining the ASD with ODT presents a reliable solution to overcome the low solubility and bioavailability problems of ATO in a simple, robust and cost effective formulation.
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Diclofenac sodium salt
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Calcium carbonate, ACS reagent, chelometric standard, 99.95-100.05% dry basis
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Magnesium stearate, technical grade
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Magnesium stearate, puriss., meets analytical specification of Ph. Eur., BP, ≥90% stearic and palmitic acid basis, ≥40% stearic acid basis (GC), 4.0-5.0% Mg basis (calc on dry sub.)
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%